siRNA Details
Gene Symbol: MICAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB048948.1 | - | 18 | 2579 | ||
AK025392.1 | - | 18 | 2346 | |||
AK296284.1 | - | 18 | 2618 | |||
BC009972.2 | - | 17 | 2076 | 4304203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
