siRNA Details
Gene Symbol: AEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209323.1 | - | 1 | 2724 | ||
AF318330.1 | - | 3 | 730 | |||
AF327352.1 | - | 2 | 721 | |||
AK022546.1 | - | 2 | 334 | |||
BC005164.1 | - | 2 | 716 | 3531883 | ||
BC014407.1 | - | 2 | 335 | 4558021 | ||
BC020988.1 | - | 3 | 738 | 3852419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
