siRNA Details
Gene Symbol: ARV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022786.1 | NP_073623.1 | 1 | 164 | ||
XM_017002106.1 | XP_016857595.1 | 1 | 159 | |||
GenBank mRNA | AF271780.1 | - | 1 | 164 | ||
AF290878.1 | - | 1 | 164 | |||
AF321442.1 | - | 1 | 198 | |||
AK026629.1 | - | 1 | 152 | |||
AK293049.1 | - | 1 | 170 | |||
BC016309.1 | - | 1 | 159 | 4047121 |
