siRNA Details
Gene Symbol: LDHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY423727.1 | - | 3 | 370 | ||
CR541714.1 | - | 3 | 339 | |||
EU794632.1 | - | 4 | 436 | |||
X02152.1 | - | 4 | 436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
