siRNA Details
Gene Symbol: FAM129B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035534.2 | NP_001030611.1 | 4 | 413 | ||
NM_022833.3 | NP_073744.2 | 4 | 625 | |||
XM_005252135.2 | XP_005252192.2 | 5 | 472 | |||
XM_011518925.1 | XP_011517227.1 | 5 | 644 | |||
GenBank mRNA | AB210016.1 | - | 4 | 363 | ||
AF151783.1 | - | 4 | 395 | |||
AF192911.1 | - | 4 | 616 | |||
AL137555.1 | - | 3 | 233 | |||
BM557756.1 | - | 587 | ||||
CN423640.1 | - | 4 | 415 |
