siRNA Details
Gene Symbol: MRPS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182640.2 | NP_872578.1 | 2, 3 | 393 | ||
GenBank mRNA | AK001451.1 | - | 2, 3 | 335 | ||
AK130078.1 | - | 2, 3 | 334 | |||
AV729381.1 | - | 2, 3 | 370 | |||
BC047784.1 | - | 1, 2 | 43 | |||
BC057240.1 | - | 2, 3 | 347 | 6495350 | ||
DT217261.1 | - | 2, 3 | 393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
