siRNA Details
Gene Symbol: MRPS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130078.1 | - | 8, 9 | 827 | ||
BC036575.1 | - | 9, 10 | 839 | |||
BC047784.1 | - | 7, 8 | 536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
