siRNA Details
Gene Symbol: CDK15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261435.1 | NP_001248364.1 | 3 | 429 | ||
NM_001261436.1 | NP_001248365.1 | 3 | 429 | |||
NM_139158.2 | NP_631897.1 | 3 | 339 | |||
XM_005246781.3 | XP_005246838.1 | 3 | 429 | |||
XM_005246782.4 | XP_005246839.1 | 4 | 676 | |||
XM_011511650.2 | XP_011509952.1 | 3 | 429 | |||
XM_011511652.1 | XP_011509954.1 | 3 | 429 | |||
XM_011511654.2 | XP_011509956.1 | 3 | 429 | |||
GenBank mRNA | AB053308.1 | - | 3 | 293 | ||
AK131512.1 | - | 3 | 429 | |||
AK292434.1 | - | 4 | 546 | |||
AK310050.1 | - | 5 | 662 | |||
AX772791.1 | - | 3 | 293 | |||
BC038807.1 | - | 3 | 387 | 5200755 | ||
BX647394.1 | - | 3 | 393 | |||
DA050598.1 | - | 3 | 429 |
