siRNA Details
Gene Symbol: ZNF649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023074.3 | NP_075562.2 | 5 | 1179 | ||
GenBank mRNA | AK000909.1 | - | 5 | 1114 | ||
AK022706.1 | - | 5 | 1177 | |||
AK225480.1 | - | 6 | 1054 | |||
AK315487.1 | - | 5 | 1141 | |||
AL713677.1 | - | 5 | 1121 | |||
BC005368.1 | - | 5 | 1138 | 3930849 |
