siRNA Details
Gene Symbol: CENPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002876.2 | NP_001002876.1 | 2 | 191 | ||
NM_001304370.1 | NP_001291299.1 | 1 | 304 | |||
NM_001304371.1 | NP_001291300.1 | 2 | 191 | |||
NM_001304372.1 | NP_001291301.1 | 1 | 304 | |||
NM_001304373.1 | NP_001291302.1 | 1 | 304 | |||
NM_024053.4 | NP_076958.1 | 2 | 191 | |||
XM_011530368.2 | XP_011528670.1 | 2 | 178 | |||
GenBank mRNA | AK123479.1 | - | 2 | 180 | ||
AL160131.1 | - | 2 | 139 | |||
BC000705.2 | - | 2 | 138 | 3349507 | ||
BC007495.2 | - | 2 | 151 | 2966541 | ||
BE789455.1 | - | 1 | 234 | |||
BE797487.1 | - | 2 | 145 | |||
BG476718.1 | - | 1 | 263 | |||
BM045658.1 | - | 1 | 279 | |||
BQ652721.1 | - | 1 | 141 | |||
BQ922403.1 | - | 2 | 124 | |||
BU556693.1 | - | 2 | 191 | |||
BX332140.2 | - | 1 | 242 | |||
BX354019.2 | - | 1 | 238 | |||
BX417558.2 | - | 1 | 243 | |||
CR456399.1 | - | 2 | 164 | |||
CR457392.1 | - | 2 | 103 | |||
HY095078.1 | - | 1 | 305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
