siRNA Details
Gene Symbol: CENPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002876.2 | NP_001002876.1 | 3, 4 | 317 | ||
NM_001304370.1 | NP_001291299.1 | 2, 3 | 430 | |||
NM_001304371.1 | NP_001291300.1 | 3, 4 | 317 | |||
NM_001304373.1 | NP_001291302.1 | 2, 3 | 430 | |||
NM_024053.4 | NP_076958.1 | 3, 4 | 317 | |||
XM_011530368.2 | XP_011528670.1 | 3, 4 | 304 | |||
GenBank mRNA | AK123479.1 | - | 3, 4 | 306 | ||
AL160131.1 | - | 3, 4 | 265 | |||
BC000705.2 | - | 3, 4 | 264 | 3349507 | ||
BC007495.2 | - | 3, 4 | 277 | 2966541 | ||
BE789455.1 | - | 2, 3 | 362 | |||
BE797487.1 | - | 3, 4 | 271 | |||
BQ652721.1 | - | 2, 3 | 267 | |||
BQ922403.1 | - | 3, 4 | 250 | |||
BX354019.2 | - | 2, 3 | 364 | |||
CR456399.1 | - | 3, 4 | 290 | |||
CR457392.1 | - | 3, 4 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
