siRNA Details
Gene Symbol: C7orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099858.1 | NP_001093328.1 | 2 | 297 | ||
NM_024054.2 | NP_076959.2 | 2 | 292 | |||
XM_005249844.3 | XP_005249901.1 | 2 | 567 | |||
XM_017012596.1 | XP_016868085.1 | 2 | 527 | |||
XM_017012597.1 | XP_016868086.1 | 2 | 1059 | |||
GenBank mRNA | AK024820.1 | - | 2 | 265 | ||
AK225194.1 | - | 2 | 271 | |||
AK298865.1 | - | 2 | 297 | |||
BC000769.2 | - | 2 | 256 | 2964418 | ||
BC001845.2 | - | 2 | 256 | 2964418 | ||
DC303669.1 | - | 2 | 277 | |||
DC366585.1 | - | 2 | 297 |
