siRNA Details
Gene Symbol: KCTD15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024076.2 | NP_076981.2 | 7 | 1035 | ||
GenBank mRNA | BC001185.1 | - | 7 | 1019 | 3502748 | |
BC009335.2 | - | 7 | 1019 | 4128019 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
