siRNA Details
Gene Symbol: FKRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039885.2 | NP_001034974.1 | 4 | 2602 | ||
NM_024301.4 | NP_077277.1 | 4 | 2550 | |||
XM_005259247.2 | XP_005259304.1 | 5 | 2663 | |||
XM_005259248.2 | XP_005259305.1 | 5 | 2653 | |||
XM_005259249.4 | XP_005259306.1 | 5 | 2595 | |||
XM_011527306.1 | XP_011525608.1 | 3 | 2399 | |||
XM_011527307.1 | XP_011525609.1 | 4 | 2448 | |||
XM_017027297.1 | XP_016882786.1 | 4 | 2769 | |||
GenBank mRNA | AJ314847.1 | - | 4 | 2538 | ||
AK022638.1 | - | 4 | 2538 | |||
AK095497.1 | - | 4 | 2589 |
