siRNA Details
Gene Symbol: CRYGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC074944.2 | - | 3 | 362 | 30915273 | |
BC074945.2 | - | 3 | 362 | 30915496 | ||
BC117384.1 | - | 3 | 362 | 40125935 | ||
BC117388.1 | - | 3 | 362 | 40125939 | ||
KT326704.1 | - | 3 | 367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
