siRNA Details
Gene Symbol: OR51B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033180.4 | NP_149420.4 | 1 | 465 | ||
GenBank mRNA | BC117461.1 | - | 1 | 465 | 40126012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
