siRNA Details
Gene Symbol: RIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206671.2 | NP_001193600.1 | 4 | 562 | ||
NM_024557.4 | NP_078833.3 | 4 | 562 | |||
NR_045405.1 | 5 | 611 | ||||
XM_006718317.3 | XP_006718380.1 | 4 | 607 | |||
XM_006718318.3 | XP_006718381.1 | 4 | 607 | |||
XM_006718321.3 | XP_006718384.1 | 4 | 607 | |||
XM_017018287.1 | XP_016873776.1 | 4 | 607 | |||
XM_017018288.1 | XP_016873777.1 | 5 | 604 | |||
XM_017018289.1 | XP_016873778.1 | 5 | 638 | |||
XM_017018290.1 | XP_016873779.1 | 4 | 607 | |||
GenBank mRNA | AK021670.1 | - | 2 | 450 | ||
AK293933.1 | - | 4 | 525 | |||
AK296505.1 | - | 5 | 576 | |||
AK315379.1 | - | 4 | 521 | |||
AY326435.1 | - | 4 | 508 | |||
BC022455.1 | - | 4 | 544 | 4792930 | ||
DC420812.1 | - | 4 | 562 | |||
KU178773.1 | - | 4 | 508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
