siRNA Details
Gene Symbol: EFCAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142857.1 | NP_001136329.1 | 3 | 446 | ||
NM_024593.3 | NP_078869.1 | 4 | 602 | |||
NR_024605.1 | 3 | 399 | ||||
XM_005251303.1 | XP_005251360.1 | 4 | 559 | |||
XM_011517589.1 | XP_011515891.1 | 4 | 559 | |||
XM_011517590.2 | XP_011515892.1 | 5 | 732 | |||
XM_011517591.2 | XP_011515893.1 | 5 | 705 | |||
XM_017013826.1 | XP_016869315.1 | 3 | 460 | |||
XM_017013827.1 | XP_016869316.1 | 6 | 855 | |||
GenBank mRNA | AK021829.1 | - | 4 | 460 | ||
AK299659.1 | - | 3 | 403 | |||
AK307101.1 | - | 3 | 389 | |||
AK316037.1 | - | 3 | 398 | |||
BC025676.1 | - | 4 | 523 | 5203436 | ||
BP375493.1 | - | 3 | 348 | |||
DA174619.1 | - | 3 | 349 | |||
DC336596.1 | - | 3 | 376 |
