siRNA Details
Gene Symbol: AKIRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136275.1 | NP_001129747.1 | 2 | 388 | ||
NM_024595.2 | NP_078871.1 | 2 | 388 | |||
GenBank mRNA | AK022728.1 | - | 2 | 387 | ||
AK092624.1 | - | 3 | 274 | |||
AK124799.1 | - | 3 | 308 | |||
AK126262.1 | - | 2 | 344 | |||
AK223276.1 | - | 2 | 344 | |||
AK303098.1 | - | 2 | 344 | |||
AY326465.1 | - | 2 | 65 | |||
BC001498.2 | - | 2 | 343 | |||
BC119745.1 | - | 2 | 305 | 40119229 | ||
BC119746.1 | - | 2 | 132 | |||
DB727813.1 | - | 2 | 409 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
