siRNA Details
Gene Symbol: AKIRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024595.2 | NP_078871.1 | 3, 4 | 630 | ||
GenBank mRNA | AK022728.1 | - | 3, 4 | 629 | ||
AK092624.1 | - | 4, 5 | 516 | |||
AK124799.1 | - | 4, 5 | 550 | |||
AK126262.1 | - | 3, 4 | 586 | |||
AK223276.1 | - | 3, 4 | 586 | |||
AK298890.1 | - | 2, 3 | 445 | |||
AY326465.1 | - | 3, 4 | 307 | |||
BC001498.2 | - | 3, 4 | 585 | |||
BC119745.1 | - | 3, 4 | 547 | 40119229 | ||
BC119746.1 | - | 3, 4 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
