siRNA Details
Gene Symbol: PPP1R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201329.1 | NP_001188258.1 | 2 | 784 | ||
NM_024607.3 | NP_078883.2 | 2 | 636 | |||
XM_006716253.3 | XP_006716316.1 | 2 | 589 | |||
XM_017013835.1 | XP_016869324.1 | 2 | 736 | |||
GenBank mRNA | AK024067.1 | - | 2 | 622 | ||
AK096119.1 | - | 2 | 716 | |||
BC043388.1 | - | 2 | 569 | 6150188 | ||
BQ431880.1 | - | 2 | 568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
