siRNA Details
Gene Symbol: NEIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256552.1 | NP_001243481.1 | 8 | 1316 | ||
NM_024608.3 | NP_078884.2 | 8 | 1392 | |||
NR_046311.1 | 9 | 1515 | ||||
XM_005254659.4 | XP_005254716.1 | 8 | 1252 | |||
XM_006720677.3 | XP_006720740.1 | 8 | 1237 | |||
XM_006720678.3 | XP_006720741.1 | 7 | 1147 | |||
XM_006720679.3 | XP_006720742.1 | 8 | 1175 | |||
XM_006720680.1 | XP_006720743.1 | 9 | 1407 | |||
XM_006720681.1 | XP_006720744.1 | 9 | 1254 | |||
XM_011522001.2 | XP_011520303.2 | 8 | 1157 | |||
XM_011522002.1 | XP_011520304.1 | 9 | 1380 | |||
XM_011522003.2 | XP_011520305.1 | 10 | 1436 | |||
XM_011522004.2 | XP_011520306.1 | 8 | 1124 | |||
XM_011522005.1 | XP_011520307.1 | 9 | 1466 | |||
XM_011522006.1 | XP_011520308.1 | 8 | 1390 | |||
XM_011522007.1 | XP_011520309.1 | 9 | 1486 | |||
XM_011522008.1 | XP_011520310.1 | 9 | 1459 | |||
XM_017022566.1 | XP_016878055.1 | 7 | 1119 | |||
XM_017022567.1 | XP_016878056.1 | 8 | 1150 | |||
GenBank mRNA | AB079068.1 | - | 7 | 886 | ||
AK026055.1 | - | 9 | 1313 | |||
AK026216.1 | - | 2 | 669 | |||
AK097008.1 | - | 9 | 1515 | |||
AK128372.1 | - | 6 | 2560 | |||
BC010876.1 | - | 8 | 1297 | 3879198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
