siRNA Details
Gene Symbol: PARP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178055.1 | NP_001171526.1 | 13 | 1118 | ||
NM_001178056.1 | NP_001171527.1 | 12 | 1094 | |||
NM_024615.3 | NP_078891.2 | 12 | 1094 | |||
XM_005248596.4 | XP_005248653.1 | 13 | 1474 | |||
XM_011543631.2 | XP_011541933.1 | 13 | 1007 | |||
XM_011543632.2 | XP_011541934.2 | 13 | 1054 | |||
XM_011543633.2 | XP_011541935.2 | 13 | 1054 | |||
XM_011543634.2 | XP_011541936.1 | 14 | 1603 | |||
XM_011543635.2 | XP_011541937.1 | 14 | 1124 | |||
XM_011543636.2 | XP_011541938.1 | 13 | 988 | |||
XM_011543640.2 | XP_011541942.1 | 10 | 878 | |||
XM_011543641.2 | XP_011541943.1 | 11 | 855 | |||
XM_011543642.2 | XP_011541944.1 | 9 | 842 | |||
XM_011543643.2 | XP_011541945.1 | 10 | 1283 | |||
XM_017009850.1 | XP_016865339.1 | 12 | 1554 | |||
XM_017009851.1 | XP_016865340.1 | 11 | 994 | |||
GenBank mRNA | AK024961.1 | - | 2 | 164 | ||
AK225188.1 | - | 2 | 164 | |||
AK289369.1 | - | 12 | 1077 | |||
AK298680.1 | - | 11 | 957 | |||
AK314466.1 | - | 13 | 1118 | |||
AL834477.1 | - | 12 | 1053 | |||
BC037386.1 | - | 10 | 952 | 4830820 | ||
BC075801.1 | - | 12 | 1053 | 5226828 |
