siRNA Details
Gene Symbol: C3orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024616.2 | NP_078892.2 | 5 | 660 | ||
XM_011513133.2 | XP_011511435.1 | 5 | 569 | |||
XM_017007181.1 | XP_016862670.1 | 5 | 1634 | |||
GenBank mRNA | AK026839.1 | - | 5 | 604 | ||
AY830714.1 | - | 5 | 606 | |||
BC017064.1 | - | 5 | 593 | 3885219 |
