siRNA Details
Gene Symbol: C3orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171747.1 | NP_001165218.1 | 2 | 317 | ||
NM_024616.2 | NP_078892.2 | 2 | 317 | |||
XM_011513133.2 | XP_011511435.1 | 2 | 226 | |||
XM_017007181.1 | XP_016862670.1 | 2 | 1291 | |||
GenBank mRNA | AK026839.1 | - | 2 | 261 | ||
AK298076.1 | - | 2 | 215 | |||
AK303588.1 | - | 2 | 317 | |||
AY830714.1 | - | 2 | 263 | |||
BC017064.1 | - | 2 | 250 | 3885219 | ||
DB159226.1 | - | 2 | 317 |
