siRNA Details
Gene Symbol: FASTKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB058703.1 | - | 6 | 981 | ||
AK025554.1 | - | 5 | 911 | |||
AK055892.1 | - | 8 | 1710 | |||
AK074302.1 | - | 8 | 1690 | |||
AK225293.1 | - | 5 | 911 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
