siRNA Details
Gene Symbol: FASTKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281476.2 | NP_001268405.1 | 12 | 2551 | ||
NM_001322046.1 | NP_001308975.1 | 13 | 3353 | |||
NM_001322048.1 | NP_001308977.1 | 12 | 2608 | |||
NM_001322049.1 | NP_001308978.1 | 11 | 2482 | |||
NM_024622.5 | NP_078898.3 | 13 | 2680 | |||
NR_104020.2 | 12 | 2547 | ||||
XM_006712750.3 | XP_006712813.1 | 12 | 2606 | |||
XM_006712751.3 | XP_006712814.1 | 9 | 2691 | |||
XM_017004911.1 | XP_016860400.1 | 13 | 3993 | |||
XM_017004912.1 | XP_016860401.1 | 8 | 2562 | |||
GenBank mRNA | AB058703.1 | - | 11 | 1862 | ||
AK025554.1 | - | 10 | 1792 | |||
AK055892.1 | - | 12 | 2462 | |||
AK074302.1 | - | 13 | 2571 | |||
AK130797.1 | - | 12 | 2471 | |||
AK225293.1 | - | 10 | 1792 | |||
AL832058.1 | - | 13 | 2563 | |||
BC032687.2 | - | 12 | 2009 | 5581443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
