siRNA Details
Gene Symbol: FASTKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281476.2 | NP_001268405.1 | 4 | 879 | ||
NM_001322046.1 | NP_001308975.1 | 4 | 1552 | |||
NM_001322048.1 | NP_001308977.1 | 3 | 807 | |||
NM_001322049.1 | NP_001308978.1 | 3 | 810 | |||
NM_024622.5 | NP_078898.3 | 4 | 879 | |||
NR_104020.2 | 4 | 879 | ||||
XM_006712750.3 | XP_006712813.1 | 3 | 805 | |||
XM_017004911.1 | XP_016860400.1 | 4 | 2192 | |||
GenBank mRNA | AB058703.1 | - | 2 | 61 | ||
AK055892.1 | - | 4 | 790 | |||
AK074302.1 | - | 4 | 770 | |||
AK130797.1 | - | 4 | 802 | |||
AL832058.1 | - | 4 | 762 | |||
BC017950.1 | - | 3 | 701 | |||
BC032687.2 | - | 3 | 208 | 5581443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
