siRNA Details
Gene Symbol: SAP30L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131062.1 | NP_001124534.1 | 3 | 976 | ||
NM_001131063.1 | NP_001124535.1 | 3 | 961 | |||
NM_024632.5 | NP_078908.1 | 4 | 1099 | |||
NR_024084.1 | 5 | 1143 | ||||
GenBank mRNA | AF085957.1 | - | 2 | 451 | ||
AI199517.1 | - | 3 | 373 | |||
AI436556.1 | - | 3 | 388 | |||
AK021588.1 | - | 4 | 1099 | |||
AY341060.1 | - | 4 | 1042 | |||
AY846876.1 | - | 4 | 451 | |||
BC009829.2 | - | 4 | 1033 | 3937610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
