siRNA Details
Gene Symbol: ZMAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135731.1 | NP_001129203.1 | 5 | 537 | ||
NM_024645.2 | NP_078921.1 | 6 | 765 | |||
GenBank mRNA | AK023904.1 | - | 6 | 765 | ||
BC019598.1 | - | 5 | 523 | 4934088 |
