siRNA Details
Gene Symbol: THAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317776.1 | NP_001304705.1 | 6 | 2718 | ||
NM_024672.5 | NP_078948.3 | 5 | 2652 | |||
XM_005263220.2 | XP_005263277.1 | 5 | 2609 | |||
XM_011532246.2 | XP_011530548.1 | 5 | 2442 | |||
XM_017008603.1 | XP_016864092.1 | 5 | 3492 | |||
XM_017008604.1 | XP_016864093.1 | 5 | 3545 | |||
XM_017008605.1 | XP_016864094.1 | 5 | 2586 | |||
XM_017008606.1 | XP_016864095.1 | 6 | 2710 | |||
XM_017008607.1 | XP_016864096.1 | 5 | 2925 | |||
XM_017008608.1 | XP_016864097.1 | 6 | 3564 | |||
XM_017008609.1 | XP_016864098.1 | 6 | 2714 | |||
XM_017008610.1 | XP_016864099.1 | 6 | 2701 | |||
GenBank mRNA | AK026973.1 | - | 3 | 1982 | ||
AK091412.1 | - | 5 | 2549 | |||
AK225333.1 | - | 3 | 1989 | |||
BC131722.1 | - | 6 | 2599 | |||
BX648134.1 | - | 6 | 2872 |
