siRNA Details
Gene Symbol: ARMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024742.2 | NP_079018.1 | 4 | 2622 | ||
GenBank mRNA | AK024430.1 | - | 4 | 2115 | ||
AY217348.1 | - | 4 | 2622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
