siRNA Details
Gene Symbol: PTCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284403.1 | NP_001271332.1 | 2 | 158 | ||
NM_001284404.1 | NP_001271333.1 | 2 | 158 | |||
NM_001284405.1 | NP_001271334.1 | 2 | 158 | |||
NM_024754.4 | NP_079030.3 | 2 | 158 | |||
XM_005248601.2 | XP_005248658.1 | 2 | 152 | |||
XM_005248603.2 | XP_005248660.1 | 2 | 144 | |||
XM_017009866.1 | XP_016865355.1 | 2 | 137 | |||
XM_017009867.1 | XP_016865356.1 | 2 | 140 | |||
XM_017009868.1 | XP_016865357.1 | 2 | 144 | |||
GenBank mRNA | AK056761.1 | - | 2 | 822 | ||
AK298750.1 | - | 2 | 149 | |||
AK303639.1 | - | 2 | 127 | |||
AK303776.1 | - | 2 | 157 | |||
BC018720.1 | - | 2 | 134 | 4552964 | ||
DA082506.1 | - | 2 | 152 | |||
DC393817.1 | - | 2 | 158 |
