siRNA Details
Gene Symbol: ZNF552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024762.3 | NP_079038.2 | 3 | 1188 | ||
XM_005259267.3 | XP_005259324.1 | 3 | 1366 | |||
GenBank mRNA | AK025256.1 | - | 1 | 492 | ||
AK097041.1 | - | 3 | 1188 | |||
AK225219.1 | - | 1 | 492 | |||
AK293445.1 | - | 3 | 1367 | |||
BC017460.1 | - | 1 | 626 | |||
BC046237.1 | - | 1 | 626 | |||
BC062982.1 | - | 1 | 626 | |||
BC082767.1 | - | 1 | 626 |
