siRNA Details
Gene Symbol: RNF122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024787.3 | NP_079063.2 | 3, 4 | 640 | ||
XM_017013846.1 | XP_016869335.1 | 3, 4 | 536 | |||
GenBank mRNA | AK022588.1 | - | 3, 4 | 637 | ||
AU124463.1 | - | 3, 4 | 638 | |||
BC093884.1 | - | 3, 4 | 258 | 7939729 | ||
BC101573.1 | - | 3, 4 | 258 | 8069079 | ||
BF847236.1 | - | 2, 3 | 165 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175136, AK077856, AK171823, BC050268, BC038399, and others.
Gene Ontology Categories:
