siRNA Details
Gene Symbol: TREML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024807.3 | NP_079083.2 | 3 | 706 | ||
XM_011514917.2 | XP_011513219.1 | 2 | 431 | |||
GenBank mRNA | AF534824.1 | - | 3 | 705 | ||
AK023755.1 | - | 3 | 705 | |||
AY358171.1 | - | 3 | 707 | |||
BC125078.1 | - | 3 | 705 | 40117710 | ||
BC125079.1 | - | 3 | 705 | 40117711 |
