siRNA Details
Gene Symbol: ZNF613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031721.3 | NP_001026891.2 | 6 | 1126 | ||
NM_024840.3 | NP_079116.2 | 6 | 1018 | |||
XM_005259269.3 | XP_005259326.1 | 5 | 1232 | |||
XM_011527333.2 | XP_011525635.1 | 6 | 1342 | |||
XM_017027315.1 | XP_016882804.1 | 5 | 1190 | |||
GenBank mRNA | AK027565.1 | - | 6 | 1018 | ||
BC057776.1 | - | 6 | 1085 | 30338140 |
