siRNA Details
Gene Symbol: CYBRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256909.1 | NP_001243838.1 | 2 | 220 | ||
NM_024843.3 | NP_079119.3 | 2 | 558 | |||
GenBank mRNA | AK027115.1 | - | 2 | 400 | ||
AK225459.1 | - | 2 | 400 | |||
AK291731.1 | - | 2 | 408 | |||
AK301485.1 | - | 2 | 220 | |||
AK316588.1 | - | 2 | 437 | |||
AL136693.1 | - | 2 | 441 | |||
BC065290.1 | - | 2 | 393 | 6063145 | ||
CX866825.1 | - | 2 | 449 | |||
DB235339.1 | - | 2 | 558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
