siRNA Details
Gene Symbol: ATAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024857.4 | NP_079133.3 | 21 | 4933 | ||
XM_011525269.2 | XP_011523571.1 | 21 | 4957 | |||
XM_011525270.2 | XP_011523572.1 | 20 | 4780 | |||
XM_011525274.2 | XP_011523576.1 | 16 | 2185 | |||
XM_017025111.1 | XP_016880600.1 | 21 | 4932 | |||
GenBank mRNA | AJ314648.2 | - | 21 | 4901 | ||
AY557611.1 | - | 21 | 4901 |
