siRNA Details
Gene Symbol: MAGIX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024859.3 | NP_079135.3 | 4 | 338 | ||
XM_005278064.4 | XP_005278121.1 | 2 | 271 | |||
XM_005278065.3 | XP_005278122.1 | 2 | 359 | |||
XM_006724560.3 | XP_006724623.1 | 4 | 422 | |||
XM_011543985.2 | XP_011542287.1 | 3 | 492 | |||
XM_011543986.2 | XP_011542288.1 | 2 | 246 | |||
XM_011543987.2 | XP_011542289.1 | 2 | 267 | |||
GenBank mRNA | AK025340.1 | - | 2 | 193 | ||
AK096058.1 | - | 2 | 1433 | |||
DA072017.1 | - | 3 | 120 | |||
DQ884401.1 | - | 1 | 114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
