siRNA Details
Gene Symbol: SLC35E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024881.4 | NP_079157.3 | 5 | 847 | ||
GenBank mRNA | AK027699.1 | - | 5 | 641 | ||
AK027850.1 | - | 3 | 258 | |||
AK075355.1 | - | 3 | 286 | |||
BC014557.2 | - | 2 | 196 | |||
BC062562.1 | - | 5 | 847 | 6598965 | ||
BX640756.1 | - | 5 | 458 |
