siRNA Details
Gene Symbol: C14orf159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102366.2 | NP_001095836.1 | 8 | 1270 | ||
NM_001102367.2 | NP_001095837.1 | 7 | 1036 | |||
NM_001102368.2 | NP_001095838.1 | 6 | 869 | |||
NM_001102369.2 | NP_001095839.1 | 8 | 1025 | |||
NM_001286470.1 | NP_001273399.1 | 9 | 1564 | |||
NM_001286471.1 | NP_001273400.1 | 6 | 1105 | |||
NM_001286472.1 | NP_001273401.1 | 8 | 1040 | |||
NM_024952.7 | NP_079228.4 | 6 | 1099 | |||
GenBank mRNA | AB210031.1 | - | 6 | 770 | ||
AK022564.1 | - | 6 | 670 | |||
AK023200.1 | - | 8 | 823 | |||
AK091394.1 | - | 7 | 459 | |||
AK097294.1 | - | 6 | 819 | |||
AK098752.1 | - | 6 | 820 | |||
AK123532.1 | - | 9 | 1207 | |||
AK308857.1 | - | 4 | 532 | |||
AY358516.1 | - | 6 | 1105 | |||
BC010614.2 | - | 7 | 985 | 4157432 | ||
BC065558.1 | - | 8 | 893 | 6138321 | ||
BX161393.1 | - | 6 | 798 | |||
BX247989.1 | - | 8 | 968 | |||
BX248294.1 | - | 4 | 471 | |||
BX248782.1 | - | 7 | 819 | |||
CR457289.1 | - | 4 | 517 |
