siRNA Details
Gene Symbol: IFT74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099222.1 | NP_001092692.1 | 17 | 1612 | ||
NM_001099223.1 | NP_001092693.1 | 17 | 1568 | |||
NM_025103.2 | NP_079379.2 | 17 | 1604 | |||
XM_011518035.2 | XP_011516337.1 | 17 | 1604 | |||
XM_011518036.2 | XP_011516338.1 | 10 | 1314 | |||
XM_017015163.1 | XP_016870652.1 | 10 | 1119 | |||
XM_017015164.1 | XP_016870653.1 | 10 | 2560 | |||
GenBank mRNA | AK023707.1 | - | 17 | 1565 | ||
AK026274.1 | - | 7 | 594 | |||
AK225570.1 | - | 17 | 1565 | |||
AY040325.1 | - | 17 | 1533 | |||
BC023634.2 | - | 17 | 1549 | |||
BC056875.1 | - | 15 | 1312 | |||
BC113524.1 | - | 16 | 1377 | |||
BC113526.1 | - | 16 | 1378 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
