siRNA Details
Gene Symbol: RUFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040451.2 | NP_001035541.1 | 2 | 603 | ||
NM_001040452.2 | NP_001035542.1 | 3 | 414 | |||
NM_025158.4 | NP_079434.3 | 3 | 589 | |||
XM_006714921.3 | XP_006714984.2 | 3 | 599 | |||
XM_017009891.1 | XP_016865380.1 | 3 | 414 | |||
XM_017009894.1 | XP_016865383.1 | 3 | 322 | |||
XM_017009895.1 | XP_016865384.1 | 2 | 394 | |||
GenBank mRNA | AB209507.1 | - | 3 | 314 | ||
AF312367.1 | - | 3 | 580 | |||
AF361055.2 | - | 3 | 562 | |||
AK075021.1 | - | 2 | 502 | |||
AK291926.1 | - | 2 | 603 | |||
BC032571.1 | - | 3 | 333 | 5243223 | ||
BP358497.1 | - | 2 | 511 | |||
DA648204.1 | - | 3 | 415 | |||
DA811890.1 | - | 2 | 502 | |||
DA925727.1 | - | 3 | 375 |
