siRNA Details
Gene Symbol: VWA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025258.2 | NP_079534.2 | 2 | 384 | ||
XM_005249427.2 | XP_005249484.1 | 2 | 384 | |||
XM_017011327.1 | XP_016866816.1 | 1 | 419 | |||
XM_017011328.1 | XP_016866817.1 | 2 | 530 | |||
GenBank mRNA | AK304058.1 | - | 2 | 360 | ||
AK304266.1 | - | 1 | 157 | |||
BC114625.1 | - | 1 | 143 | |||
DC323829.1 | - | 2 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
