siRNA Details
Gene Symbol: TMEM177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105198.1 | NP_001098668.1 | 2 | 819 | ||
NM_001105199.1 | NP_001098669.1 | 2 | 427 | |||
NM_030577.2 | NP_085054.2 | 2 | 478 | |||
GenBank mRNA | AK096890.1 | - | 2 | 424 | ||
AK130088.1 | - | 2 | 427 | |||
AK299078.1 | - | 2 | 424 | |||
BC004404.1 | - | 2 | 466 | 3637432 | ||
BG280893.1 | - | 2 | 421 | |||
BG776153.1 | - | 2 | 449 |
