siRNA Details
Gene Symbol: ASXL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030632.2 | NP_085135.1 | 12 | 3794 | ||
XM_005258356.1 | XP_005258413.1 | 13 | 3767 | |||
XM_011526205.2 | XP_011524507.1 | 13 | 10557 | |||
XM_011526206.2 | XP_011524508.1 | 13 | 7322 | |||
XM_011526209.1 | XP_011524511.1 | 14 | 3951 | |||
XM_011526210.1 | XP_011524512.1 | 14 | 4008 | |||
XM_011526212.1 | XP_011524514.1 | 13 | 3912 | |||
XM_011526213.1 | XP_011524515.1 | 13 | 4005 | |||
XM_017026012.1 | XP_016881501.1 | 13 | 7169 | |||
XM_017026013.1 | XP_016881502.1 | 14 | 7531 | |||
XM_017026014.1 | XP_016881503.1 | 14 | 3968 | |||
GenBank mRNA | AB051500.2 | - | 2 | 1953 |
