siRNA Details
Gene Symbol: OR2C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198074.4 | NP_932340.3 | 2 | 1232 | ||
GenBank mRNA | BC030717.1 | - | 2 | 1230 | 4860551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
