siRNA Details
Gene Symbol: VWA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045104.1 | 2 | 463 | |||
NR_045105.2 | 2 | 717 | ||||
NR_045106.2 | 2 | 624 | ||||
NR_045901.1 | 2 | 463 | ||||
XM_006720708.3 | XP_006720771.1 | 2 | 399 | |||
GenBank mRNA | AK027475.1 | - | 2 | 305 | ||
AK296134.1 | - | 2 | 690 | |||
AK301222.1 | - | 2 | 416 | |||
AK310857.1 | - | 2 | 309 | |||
AL136662.1 | - | 2 | 545 | |||
BC007991.2 | - | 2 | 290 | 3140504 | ||
BC037823.1 | - | 1 | 1015 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
