siRNA Details
Gene Symbol: VWA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136043.2 | NP_001129515.1 | 5 | 667 | ||
NM_001207058.1 | NP_001193987.1 | 5 | 646 | |||
NM_001207059.1 | NP_001193988.1 | 4 | 526 | |||
NR_045104.1 | 5 | 810 | ||||
NR_045105.2 | 5 | 1064 | ||||
NR_045106.2 | 5 | 971 | ||||
NR_045901.1 | 4 | 702 | ||||
XM_006720708.3 | XP_006720771.1 | 5 | 746 | |||
XM_006720709.3 | XP_006720772.1 | 5 | 811 | |||
XM_006720711.2 | XP_006720774.1 | 4 | 670 | |||
XM_011522101.2 | XP_011520403.1 | 5 | 492 | |||
XM_017022644.1 | XP_016878133.1 | 5 | 891 | |||
GenBank mRNA | AK027475.1 | - | 5 | 652 | ||
AK293207.1 | - | 4 | 479 | |||
AK296134.1 | - | 5 | 1037 | |||
AK301033.1 | - | 5 | 509 | |||
AK301222.1 | - | 4 | 655 | |||
AK301742.1 | - | 5 | 488 | |||
AL136662.1 | - | 5 | 892 | |||
BC007991.2 | - | 5 | 637 | 3140504 | ||
DC298728.1 | - | 4 | 479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
